missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Reach pH Electrodes
A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.
£243.00 - £423.00
Specifications
Junction | Ceramic |
---|---|
Material | Glass |
pH Range | 0 to 14 |
For Use With (Application) | Accurate measurement in a wide range of sample types |
Membrane | HA Glass |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10523375
|
METTLER TOLEDO™
51343062 |
InLab™ Reach Pt1000-425 | Combination Electrode; Integrated Temperature Probe (Pt-1000) | Multi-Pin | Multi-Pin to BNC (30281896) |
£373.00
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||
10049392
|
METTLER TOLEDO™
51343061 |
InLab™ Reach Pro-425 | Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ) | Multi-Pin | Multi-Pin to BNC (30281896) |
£423.00
Each |
Estimated Shipment: 19-04-2024
Log in to see stock. |
||
15529601
|
METTLER TOLEDO™
30244733 |
InLab™ Reach 225 | Combination Electrode | S7 | S7 to BNC (30281915) |
£243.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
15569601
|
METTLER TOLEDO™
30248120 |
InLab™ Reach 425 | Combination Electrode | S7 | S7 to BNC (30281915) |
£282.00
Each |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||
15579601
|
METTLER TOLEDO™
30248826 |
InLab™ Reach Pro-225 | Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ) | Multi-Pin | Multi-Pin to BNC (30281896) |
£347.00
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
||
15599601
|
METTLER TOLEDO™
30248828 |
InLab™ Reach Pt1000-225 | Combination Electrode; Integrated Temperature Probe (Pt-1000) | Multi-Pin | Multi-Pin to BNC (30281896) |
£337.00
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Pilot reactor
- Tank and Barrel
- Large samples
- Deep vessels
Accessories & Care
- Ceramic junction
- HA glass membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Ceramic | |
0 to 14 | |
HA Glass |
Glass | |
Accurate measurement in a wide range of sample types | |
0°C to 100°C |