Learn More
Mettler Toledo™ LocalCAN Interface Cables
For connecting balances with peripherals
Brand:  Mettler Toledo™ 00229050
Code : 50
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12734732
  • £238.43 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Use with discontinued AG, PR, and SR Series balances.
  • One end has 4-pin connector that connects to LocalCAN universal interface of balance
  • Other end has one of several connectors as indicated in chart
  • All are 5 ft. long (1.5m)
AB, AG, PR, and SR series balances and HR83P Moisture Analyzer must be equipped with accessory LC-B LocalCAN universal interface (01-911-153B, available online) to use the interface cables. PG-S, XS, XP, and AX Series balances must be equipped with optional LocalCan interface. (PG-S requires LC-G interface 01-918-91; XS/XP requires 09-320-113; AX requires 09-320-114.)
Specifications
Specifications
AG, PR, SR balance
DB25
60 in.
Connecting printer or PC with RS232C
Videos