Learn More
Mettler Toledo™ ISA Solutions
Solutions for perfectION™ combined ion selective electrodes
Brand: Mettler Toledo™ 51344766
Code : 59
Additional Details : Weight : 0.59000kg
Description
Solutions (ionic-strength adjustment buffers)
- Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
- ISA for calcium electrode, 475mL
- ISA for potassium electrode, 475mL
- ISA for nitrate electrode, 475mL
- Nitrate ISS (for suppressing interference), 475mL
- Fluoride TISAB II with CDTA, 3790mL
- Fluoride TISAB III with CDTA (concentrate), 475mL
Specifications
perfectION TISAB III with CDTA | |
Fluoride TISAB III with CDTA | |
475 mL |
ISA Solution | |
475 mL |
Safety and Handling
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P321-Specific treatment (see on this label).