Learn More
METTLER TOLEDO™ InLab™ Mono Half-Cell pH Measuring Electrodes
A half-cell pH measuring electrode featuring a glass body that is suited for high temperature and high pH samples in which the lifetime of the electrode is significantly shorter than that of a standard reference electrode.
Brand: METTLER TOLEDO™ 51343196
Code : 59
Additional Details : Weight : 0.07000kg
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- High temperature samples
- High pH samples
- Corrosive acids and bases
- Drinking water
Accessories & Care
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 to BNC (30281915) | |
0 to 12 | |
Half-Cell Measuring Electrode; A41 Thick-Walled Membrane Glass | |
InLab™ Mono Plus | |
0°C to 130°C | |
120 mm | |
12 mm |
Glass | |
0°C to 130°C | |
Electrolyte hose coupling, Electrolyte bridge | |
32°F to 266°F | |
4.7 in. | |
0.47 in. |