missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy Plus™ FEP30 Conductivity Meter

Economical and reliable solution for easy conductivity measurement

Brand:  Mettler Toledo™ 51302935

Code : 59

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12986387

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

FEP30-Basic
±0.5%
0 to 19.99psu
1
0.1°C
LCD
Mini-DIN
9VDC
0.1μS/cm to 199.9mS/cm
0.1mg/L to 199.9g/L
Conductivity
0° to 100°C
±0.3°C
99
RS232
100/240V
Videos
Documentation