missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveEasy™ Meters

For use with FiveEasy pH meters.

Brand:  Mettler Toledo™ 51302952

UNSPSC : 41115600

Code : 59

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12611296

  • £13.40 / Pack of 2


Estimated Shipment
12-09-2017
Add to basket

Description

Description

  • Covers electrode arm opening on instrument
  • Two pieces
Specifications

Specifications

pH meter
pH
Rubber
Electrode Arm Socket Cap
Documentation