missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveEasy™ Meters

For use with FiveEasy pH meters.

Brand:  Mettler Toledo™ 51302951

UNSPSC : 41115604

Code : 59

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 10398203

  • £60.45 / Each


In Stock 1
Add to basket

Description

Description

  • Can be attached to the left or right-hand side of the instrument
Specifications

Specifications

pH meter
pH
Electrode Arm
Documentation