missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ F1 Premium Wire Weights with Certificate

Individual weights

Brand:  Mettler Toledo™ 00159467

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12763155

  • £50.48 / Each


Estimated Shipment
23-02-2018
Add to basket

Specifications

Specifications

Wire weight
OIML Class F1
50mg
50mg
Plastic box
1
With certificate, ISO17025, ISO9001, ISO14001 environmental standard
Stainless Steel
8kg/dm3 at 20°C
Triangle
Each
Videos