missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ EL20 Benchtop pH Meter for Teaching and Learning

Compact instrument tailored for training courses in colleges and universities

Brand:  Mettler Toledo™ 51302930

Code : 59

Additional Details : Weight : 1.30000kg

 View more versions of this product

Product Code. 11947991

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

±0.5°C
±0.01
0 to 14pH
pH, mV, Temperature
Meter, operating instructions, quick guide, power adapter
20cm
2mm Banana socket for analog output
0.1°C
0° to 100°C
0.6kg
LCD
0 to 14
±1mV
1 or 2
120/240V, 50/60Hz, 9VDC
50/60Hz adapter for 9V DC input
BNC for pH combination or half cell, 2mm banana socket for separate reference half cell, Cinch/RCA jack for ATC probe (NTC 30kilohm)
ABS/PC Reinforced Window, Membrane Keypad
NTC
1mV
120/240V
17.5cm
0 to 1999mV
0.01
Alternative Products