missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Replacement Tubing for DL31 Karl Fischer™ Titrators
Replacement parts for use with DL31 Karl Fischer titrators.
Brand: Mettler Toledo™ 51107481
Code : 04
Additional Details : Weight : 0.25000kg
Description
For use with DL31 Karl Fischer Titrators.Specifications
Solvent Delivery/Drain Tube with Screw Fitting | |
Titration | |
Solvent Dispensing Tubes, Suction Pipe | |
Mettler Toledo V20/V30 KF Titrators |