missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Detachable Cables for S7 Screw Head Electrodes

Connects electrodes with an S7 screw head to meters with specified connector. Mettler Toledo Detachable Cables can be used with meters that have a BNC, 2mm Banana or US Standard connector.

Brand:  Mettler Toledo™ 52300014

Code : 59

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 10085611

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Detachable cables that attach to electrodes with an S7 screw head
  • Length 1.2m (3.9 ft.)

Specifications

Specifications

US Standard
S7 Screw head electrode with US standard connector
Cable
1.2m
Grey
Videos