missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Density Kits

For use with XS/XP balances. Mettler Toledo Density Kits help to determine the density of solids and liquids with top-loading electronic balances.

Brand:  Mettler Toledo™ 11106706

Code : 50

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 10567945

  • £676.62 / 1 set


Estimated Shipment
14-05-2018
Add to basket

Description

Description

Solids Kit

  • 200mL beaker
  • Thermometer
  • Bridge
  • Sample Holder (2)
  • Bracket
  • Bottle of wetting agent, 25mL
Liquids Kit
  • Sinker

 

Specifications

Specifications

XS/XP analytical
Solids kit includes 200mL beaker, thermometer, bridge, sample holder (2), bracket, bottle of wetting agent (25mL)
Videos
Documentation