missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Balance Printer Accessories: Printer Paper
Printer paper for use with Mettler Toledo™ balances.
Brand: Mettler Toledo™ 72456
Additional Details : Weight : 0.28620kg
Description
- Color: White
- Five rolls per pack
- For use with Mettler Toledo balances: LC-P45, LC-P43, HA-P43, HR-73P
Specifications
LC-P45, LC-P43, HA-P43, HR-73P Printer | |
5 Rolls/Pk. | |
Printer Paper |