missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveGo™ pH Meters

For use with FiveGo pH meters.

Brand:  Mettler Toledo™ 51302960

Code : 59

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11997741

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Attaches electrode to side of meter
Specifications

Specifications

pH meter
pH
Electrode Clip
Videos
Documentation