missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveGo™ pH Meters

For use with FiveGo pH meters.

Brand:  Mettler Toledo™ 51302966

UNSPSC : 41115600

Code : 59

Additional Details : Weight : 0.65000kg

 View more versions of this product

Product Code. 11957751

  • £46.10 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Description

For use with FiveGo pH meters.
Description & Specifications

Specifications

FiveGo™ pH meter
Carry Bag
pH
Documentation