missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveGo™ pH Meters

For use with FiveGo pH meters.

Brand:  Mettler Toledo™ 51302962

UNSPSC : 26111723

Code : 59

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11927751

  • £32.62 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

For use with FiveGo pH meters.
Specifications

Specifications

pH meter
White
Battery Cover
O-ring
pH
Documentation