Learn More
METTL6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557185
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MASLQRKGLQ ARILTSEEEE KLKRDQTLVS DFKQQKLEQE AQKNWDLFYK RNSTNFFKDR HWTTREFEEL RSCREFEDQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.
S-adenosyl-L-methionine-dependent methyltransferase that mediates N(3)-methylcytidine modification of residue 32 of the tRNA anticodon loop of tRNA(Ser). [UniProt]Specifications
METTL6 | |
Polyclonal | |
Unconjugated | |
METTL6 | |
1600013P15Rik; AI467521; AU020711; methyltransferase like 6; methyltransferase-like protein 6; Mettl6; tRNA N(3)-methylcytidine methyltransferase METTL6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131965 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TCB7 | |
METTL6 | |
Recombinant protein corresponding to Human METTL6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only