missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Slanted Label Holders for Wire Shelving

Snap-on plastic holders provide easy identification of shelf contents

Brand:  Metro™ 9990SL2

Additional Details : Weight : 0.03200kg

 View more versions of this product

Product Code. 15966781

Quantity Price
1 £12.29 / Each
10 £11.20 / Each
Estimated Shipment: 13-04-2022

Add to basket


Description

Description

  • Gray solid plastic holders put 1.25 in. (32 mm) labels on slant for easier viewing
  • Labels not included
Specifications

Specifications

Gray
19 x 1.25 in.
24 in.
Plastic
-18 to 120.2°F
Metro™ Super Erecta™, Super Adjustable™ 2, qwikSLOT™, Drop Mat™ wire shelving, cantilever shelves
48 x 3.2 cm
61 cm
Slanted
-28 to 49°C
Videos
SDS
Documents
Certificates
Special Offers

Special Offers