missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax Shelf Posts

Compatible with MetroMax i™ and MetroMax Q™ shelves

Brand:  Metro™ MX86P

Additional Details : Weight : 0.90400kg

 View more versions of this product

Product Code. 15977211

  • £79.55 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

Grooves located every 1 in. for locking shelf wedges in place. Post height stamped to face of post for easy shelf positioning.

  • Corrosion-proof polymer posts with built-in Microban™ antimicrobial product protection
  • Stationary posts include an adjustable leveling foot to compensate for uneven floors
  • Leveling foot can be adjusted 1 in. (25mm)
  • Mobile posts accept optional Metro Stem Casters, available in a variety of materials to meet your specific needs

Specifications

Specifications

Stationary Post with Leveling Foot
86 in.
Polymer
218.4 cm
Metro™ MetroMax i™ and Q™ shelving
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers