missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf

The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.

Brand:  Metro™ LTM60XS3

Additional Details : Weight : 46.08800kg

 View more versions of this product

Product Code. 15967081

  • £1632.00 / Each

Estimated Shipment: 31-01-2022
Add to basket

Description

Description

  • Table features smooth surfaces that are easy to clean
  • Mobile units can be easily moved to clean behind table area
  • Available top materials include Type 304 stainless and TRESPA phenolic resin (black or gray)
  • Wide range of accessories and caster options available

Order Info

Casters must be ordered separately.

Specifications

Specifications

59.75 x 30 x 39.5 in. (151.7 x 76.2 x 100.3 cm)
39.5 in.
Backsplash with solid Metro™ MetroMax i™ shelf, adaptor insert
151.7 cm
30 in.
600 lb.
273 kg
Ideal for lab
100.3 cm
59.75 in.
Polymer
76.2 cm
Mobile Table
Videos
SDS
Documents
Certificates
Special Offers

Special Offers