missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Double-Deep qwikTRAK™ Stationary End Unit Kits

Connects end units to tracks and anchors tracks to floor

Brand:  Metro™ LBTEX3

Additional Details : Weight : 26.48800kg

 View more versions of this product

Product Code. 15946981

Quantity Price
1 £1754.40 / Each
3 £1598.96 / Each
Estimated Shipment: 18-03-2022

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
  • Kit includes 74 in. (188cm) high stationary posts and connecting hardware

Order Info

Order one kit per double-deep system. Shelves are sold separately.

Specifications

Specifications

Heavy Polymer
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor
19.2 kg
Stationary end unit kit
Metro™ MetroMax i™ and Q™ shelving system
42.3 lb.
Polymer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers