missing translation for 'onlineSavingsMsg'
Learn More

Methysergide maleate, Tocris Bioscience™

5-HT1/5-HT2 antagonist

Brand:  Tocris (Bio-Techne) 106450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13448423

  • £786.45 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mixed 5-HT1/5-HT2 receptor antagonist.
Specifications

Chemical Identifiers

129-49-7
469.538
methysergide maleate, methysergide dimaleate, deseril-retard, desernil bismaleate, methysergide hydrogen maleate, unii-2u7h1466gh, methysergide bimaleate, methysergide maleate usp, methysergide maleate salt, 1-methyl-d-lysergic acid butanolamide dimaleate
(6aR,9R)-N-[(2S)-1-hydroxybutan-2-yl]-4,7-dimethyl-6,6a,8,9-tetrahydroindolo[4,3-fg]quinoline-9-carboxamide;(Z)-but-2-enedioic acid
C25H31N3O6
LWYXFDXUMVEZKS-ZVFOLQIPSA-N
5281073
CCC(CO)NC(=O)C1CN(C2CC3=CN(C4=CC=CC(=C34)C2=C1)C)C.C(=CC(=O)O)C(=O)O

Specifications

50mg
129-49-7
methysergide maleate, methysergide dimaleate, deseril-retard, desernil bismaleate, methysergide hydrogen maleate, unii-2u7h1466gh, methysergide bimaleate, methysergide maleate usp, methysergide maleate salt, 1-methyl-d-lysergic acid butanolamide dimaleate
CCC(CO)NC(=O)C1CN(C2CC3=CN(C4=CC=CC(=C34)C2=C1)C)C.C(=CC(=O)O)C(=O)O
469.538
469.54
Methysergide maleate
C25H31N3O6
LWYXFDXUMVEZKS-ZVFOLQIPSA-N
(6aR,9R)-N-[(2S)-1-hydroxybutan-2-yl]-4,7-dimethyl-6,6a,8,9-tetrahydroindolo[4,3-fg]quinoline-9-carboxamide;(Z)-but-2-enedioic acid
5281073
>99%
Videos
Documents