missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyltriphenylphosphonium chloride, 97%

Brand:  Alfa Aesar™ H56851.03

Code : NEW

Additional Details : CAS Number : 1031-15-8 Weight : 0.00100kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15403398

  • £15.17 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1031-15-8
312.777
QRPRIOOKPZSVFN-UHFFFAOYSA-M
9879809
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
C19H18ClP
MFCD00797851
methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
methyl(triphenyl)phosphanium;chloride

Specifications

Methyltriphenylphosphonium chloride
221°C (decomposition)
1031-15-8
CH3P(C6H5)3Cl
UN3077
Soluble in methanol.
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
312.777
312.78
Hygroscopic
1g
C19H18ClP
MFCD00797851
methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
QRPRIOOKPZSVFN-UHFFFAOYSA-M
methyl(triphenyl)phosphanium;chloride
9879809
97%
Videos
SDS Documents
Certificates