missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenoxyphosphonium iodide, 95%, Thermo Scientific Chemicals

Catalog Number 10015443
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17579-99-6
C19H18IO3P
452.22
MFCD00011911
VKTOBGBZBCELGC-UHFFFAOYSA-M
methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
2735090
methyl(triphenoxy)phosphanium;iodide
C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17579-99-6
C19H18IO3P
452.22
MFCD00011911
VKTOBGBZBCELGC-UHFFFAOYSA-M
methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
2735090
methyl(triphenoxy)phosphanium;iodide
C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 141.0°C to 146.0°C
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 50 g
Beilstein 06, 177
Fieser 01,1249; 02,446; 04,557; 06,649; 08,354
Solubility Information Solubility in water: negligible. Other solubilities: soluble in acetone,diethyl ether
Formula Weight 452.22
Percent Purity 95%
Chemical Name or Material Methyltriphenoxyphosphonium iodide
Show More Show Less
compliance-icons
Product Identifier
  • Methyltriphenoxyphosphonium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'