missing translation for 'onlineSavingsMsg'
Learn More

Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4259048
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
1185-55-3
C4H12O3Si
136.22
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
14456
trimethoxy(methyl)silane
CO[Si](C)(OC)OC
This item is not returnable. View return policy
1185-55-3
C4H12O3Si
136.22
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
14456
trimethoxy(methyl)silane
CO[Si](C)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 102°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3Si(OCH3)3
Refractive Index 1.3695 to 1.3715
Quantity 100 mL
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes
Viscosity 0.5 mPa.s (20°C)
Formula Weight 136.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyltrimethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'