missing translation for 'onlineSavingsMsg'
Learn More

Acros Organics Methylsulfuric acid sodium salt, 99%, pract., Acros Organics

Brand:  Acros Organics 414900250

Methylsulfuric acid sodium salt, 99%, pract., Acros Organics

Code : 29

Additional Details : CAS Number : 512-42-5 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 1 Pack group : II

Product Code. 10559282

  • £56.10 / 25g


Call for latest availability
+44 (0)1509 555 500
Add to basket