missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfamic acid, 98%, ACROS Organics™

Brand:  Acros Organics 314670050

Code : 29

Additional Details : CAS Number : 4112-03-2 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10791161

  • £24.80 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Methylsulfamic acid
100.0
98%
CH3NHSO3H
MFCD00010870
97.5
≥97.5%
CH5NO3S
111.12

Specifications

Methylsulfamic acid
97.5
Authentic
Crystals or Crystalline Powder
98%
CH3NHSO3H
MFCD00010870
White
5g
100.0
Glass bottle
≥97.5%
CH5NO3S
111.12
189° to 193°C
Videos
SDS