missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number p-7051459
Click to view available options
Quantity:
1 mL
10 mL
50 mL
This item is not returnable. View return policy
1184-57-2
MFCD00013924
This item is not returnable. View return policy
1184-57-2
MFCD00013924

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 1 mL
Linear Formula CH3HgOH
UN Number UN2024
Solubility Information Slightly soluble in water.
Formula Weight 232.63
Chemical Name or Material Methylmercury(II) hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'