Learn More
Methyllycaconitine citrate, Tocris Bioscience™
Selective α7 neuronal nAChR antagonist
Brand:  Tocris (Bio-Techne) 1029/25
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 25mg
 View more versions of this product
Product Code. 13468333
  • £367.50 / 25mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Potent antagonist for α7-containing neuronal nicotinic receptors (Ki = 1.4 nM). Interacts with α4β2 and α6β2 receptors at concentrations > 40 nM. Attenuates METH-induced neurotoxicity in mouse striatum in vivo.
Specifications
Chemical Identifiers
351344-10-0
874.934
methyllycaconitine citrate
CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
C43H58N2O17
INBLZNJHDLEWPS-OXVGBIBJSA-N
124081005
Specifications
25mg
351344-10-0
methyllycaconitine citrate
CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
124081005
Solid
Methyllycaconitine citrate
C43H58N2O17
INBLZNJHDLEWPS-OXVGBIBJSA-N
874.934
874.93
>98%