missing translation for 'onlineSavingsMsg'
Learn More

Methyllithium, 3% solution in 2-MeTHF/cumene, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 445841000

Code : 29

Additional Details : CAS Number : 917-54-4 Weight : 0.13670kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I

Quantity 100mL
Packaging AcroSeal™ Glass bottle

 View more versions of this product

Product Code. 11996151

  • £33.60 / 100mL


EU Stock 3
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Methyllithium
2.7 to 3.4% (as MeLi) (Total base)
CH3Li
96-47-9
CH3Li
21.96

Specifications

Methyllithium
0.8670g/mL
0.867
2.7 to 3.4% (as MeLi) (Total base)
CH3Li
Clear colorless
−11°C
96-47-9
AcroSeal™ Glass bottle
Liquid
CH3Li
21.96
100mL
Videos
SDS