missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyllithium, 3% solution in 2-MeTHF/cumene, AcroSeal™, Thermo Scientific™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | CH3Li |
Flash Point | −11°C |
Formula Weight | 21.96 |
Percent Purity | 2.7 to 3.4% (as MeLi) (Total base) |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.867 |
Density | 0.8670g/mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'