missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyllithium, 2.2M (6wt%) in diethyl ether with LiBr, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless through Yellow |
Physical Form | Solution |
Quantity | 100 mL |
Linear Formula | CH3Li |
Flash Point | −28°C |
Solubility Information | Solubility in water: vigorous reaction |
Formula Weight | 21.98 |
Percent Purity | 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3) |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.81 |
Show More |
Safety and Handling
Product Identifier
- Methyllithium, 2.2M (6wt%) in diethyl ether with lithiumbromide
- Danger
- Flammable liquid Category 1
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Skin sensitiser Category 1
- H224-Extremely flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H336-May cause drowsiness or dizziness.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Lithium bromide (LiBr), Lithium, methyl-, Ethyl ether
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'