missing translation for 'onlineSavingsMsg'
Learn More

Methylguanidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7041725
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
22661-87-6
C2H8ClN3
109.557
MFCD00012576
VJQCNCOGZPSOQZ-UHFFFAOYSA-N
1-methylguanidine hydrochloride, methylguanidine hydrochloride, methylguanidine hcl, n-methylguanidine hydrochloride, 1-methylguanidine hcl, guanidine, methyl-, hydrochloride, 2-methylguanidine hydrochloride, 1-methylguanidine hydrochloride 1:x, guanidine, methyl-, monohydrochloride, methyl guanidine hcl
146724
2-methylguanidine;hydrochloride
CN=C(N)N.Cl
This item is not returnable. View return policy
22661-87-6
C2H8ClN3
109.557
MFCD00012576
VJQCNCOGZPSOQZ-UHFFFAOYSA-N
1-methylguanidine hydrochloride, methylguanidine hydrochloride, methylguanidine hcl, n-methylguanidine hydrochloride, 1-methylguanidine hcl, guanidine, methyl-, hydrochloride, 2-methylguanidine hydrochloride, 1-methylguanidine hydrochloride 1:x, guanidine, methyl-, monohydrochloride, methyl guanidine hcl
146724
2-methylguanidine;hydrochloride
CN=C(N)N.Cl

Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

Solubility
Soluble in water (50 mg/ml - clear, colorless to light yellow solution).

Notes
Hygroscopic, store away form water/moisture. Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml- clear,colorless to light yellow solution).
Formula Weight 109.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methylguanidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'