missing translation for 'onlineSavingsMsg'
Learn More

Methyl trimethylacetate, 99%, Thermo Scientific Chemicals

Catalog Number p-3758503
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
598-98-1
C6H12O2
116.16
MFCD00008843
CNMFHDIDIMZHKY-UHFFFAOYSA-N
methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
69027
methyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC
This item is not returnable. View return policy
598-98-1
C6H12O2
116.16
MFCD00008843
CNMFHDIDIMZHKY-UHFFFAOYSA-N
methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
69027
methyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70°C
Color Colorless
Density 0.8700g/mL
Boiling Point 101°C
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCO2CH3
Refractive Index 1.3890 to 1.391
Quantity 100 mL
Beilstein 02, 320
Specific Gravity 0.87
Viscosity 30 mPa.s (25°C)
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl trimethylacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl trimethylacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'