missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog Number p-4229312
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle in Tin Can
Glass bottle
This item is not returnable. View return policy
333-27-7
C2H3F3O3S
164.10
MFCD00000409
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
9526
methyl trifluoromethanesulfonate
COS(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
333-27-7
C2H3F3O3S
164.10
MFCD00000409
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
9526
methyl trifluoromethanesulfonate
COS(=O)(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.4500g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle in Tin Can
Linear Formula CF3SO3CH3
Refractive Index 1.3250 to 1.3270 (20°C, 589nm)
Quantity 25 g
Fieser 06,406; 12,329; 13,327; 17,191
Formula Weight 164.11
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material Methyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'