missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Thioglycolate, 95%, Thermo Scientific Chemicals
15.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -24°C |
Color | Colorless |
Density | 1.1870g/mL |
Boiling Point | 156°C to 159°C (10.0 mmHg) |
Flash Point | 56°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (Iodimetry) |
Packaging | Glass bottle |
Linear Formula | HSCH2CO2CH3 |
Refractive Index | 1.4640 to 1.467 |
Show More |
Safety and Handling
Product Identifier
- Methyl thioglycolate
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'