missing translation for 'onlineSavingsMsg'
Learn More

Methylergometrine maleate, Tocris Bioscience™

Active metabolite of methysergide (Cat. No. 1064)

Brand:  Tocris (Bio-Techne) 0549/250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 250mg

 View more versions of this product

Product Code. 13437713

  • £388.71 / 250mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Active metabolite of methysergide (Cat. No. 1064); uterotonic agent and oxytocic.
Specifications

Chemical Identifiers

57432-61-8
455.511
methylergonovine maleate, methylergometrine maleate, methergine, erezingen, methylergobasine maleate, usaf uctl-8, unii-ir84jpz1rk, methylergonovine maleate salt, maleic acid, methyl ergonovine, methergine maleate salt
(6aR,9R)-N-[(2S)-1-hydroxybutan-2-yl]-7-methyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide;(Z)-but-2-enedioic acid
C24H29N3O6
NOFOWWRHEPHDCY-DAUURJMHSA-N
5281072
CCC(CO)NC(=O)C1CN(C2CC3=CNC4=CC=CC(=C34)C2=C1)C.C(=CC(=O)O)C(=O)O

Specifications

Methylergometrine maleate
57432-61-8
methylergonovine maleate, methylergometrine maleate, methergine, erezingen, methylergobasine maleate, usaf uctl-8, unii-ir84jpz1rk, methylergonovine maleate salt, maleic acid, methyl ergonovine, methergine maleate salt
CCC(CO)NC(=O)C1CN(C2CC3=CNC4=CC=CC(=C34)C2=C1)C.C(=CC(=O)O)C(=O)O
455.511
455.51
250mg
C24H29N3O6
NOFOWWRHEPHDCY-DAUURJMHSA-N
(6aR,9R)-N-[(2S)-1-hydroxybutan-2-yl]-7-methyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide;(Z)-but-2-enedioic acid
5281072
>98%
Videos