missing translation for 'onlineSavingsMsg'
Learn More

Methyl propiolate, 99%, Thermo Scientific Chemicals

Catalog Number 10478320
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
922-67-8
C4H4O2
84.07
MFCD00008572
IMAKHNTVDGLIRY-UHFFFAOYSA-N
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
13536
methyl prop-2-ynoate
COC(=O)C#C
This item is not returnable. View return policy
922-67-8
C4H4O2
84.07
MFCD00008572
IMAKHNTVDGLIRY-UHFFFAOYSA-N
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
13536
methyl prop-2-ynoate
COC(=O)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9400g/mL
Boiling Point 98°C to 101°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HC∽CCO2CH3
Refractive Index 1.4070 to 1.409
Quantity 50 g
Beilstein 02, I, 208
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl propiolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl propiolate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'