missing translation for 'onlineSavingsMsg'
Learn More

Methyl picolinate, 98%, Thermo Scientific Chemicals

Catalog Number 10687410
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2459-07-6
C7H7NO2
137.14
NMMIHXMBOZYNET-UHFFFAOYSA-N
COC(=O)C1=CC=CC=N1
This item is not returnable. View return policy
2459-07-6
C7H7NO2
137.14
NMMIHXMBOZYNET-UHFFFAOYSA-N
COC(=O)C1=CC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14°C to 16°C
Color White
Density 1.1370g/mL
Boiling Point 95°C (1.0 hPa)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.137
Synonym methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate
IUPAC Name methyl pyridine-2-carboxylate
PubChem CID 75569
Formula Weight 137.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl picolinate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl picolinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'