missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfoxide, 98%, Thermo Scientific Chemicals

Catalog Number p-4089552
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
934-72-5
C8H10OS
154.23
MFCD00075175
FEVALTJSQBFLEU-UHFFFAOYNA-N
1-methyl-4-methylsulfinyl benzene, methyl 4-methylphenylsulfoxide, methyl p-tolyl sulfoxide, 1-methanesulfinyl-4-methylbenzene, methyl p-methylphenylsulfoxide, 1-methyl-4-methylsulphinyl benzene, benzene, 1-methyl-4-methylsulfinyl, acmc-20aotg, acmc-20apjq, methyl p-tolylsulfoxide
136743
1-methyl-4-methylsulfinylbenzene
CC1=CC=C(C=C1)S(C)=O
This item is not returnable. View return policy
934-72-5
C8H10OS
154.23
MFCD00075175
FEVALTJSQBFLEU-UHFFFAOYNA-N
1-methyl-4-methylsulfinyl benzene, methyl 4-methylphenylsulfoxide, methyl p-tolyl sulfoxide, 1-methanesulfinyl-4-methylbenzene, methyl p-methylphenylsulfoxide, 1-methyl-4-methylsulphinyl benzene, benzene, 1-methyl-4-methylsulfinyl, acmc-20aotg, acmc-20apjq, methyl p-tolylsulfoxide
136743
1-methyl-4-methylsulfinylbenzene
CC1=CC=C(C=C1)S(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 46°C
Boiling Point 113°C to 114°C (2.2 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material Methyl p-tolyl sulfoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'