missing translation for 'onlineSavingsMsg'
Learn More

Methyl oxalyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3736992
Click to view available options
Quantity:
25 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
5781-53-3
C3H3ClO3
122.51
MFCD00000705
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
79846
methyl 2-chloro-2-oxoacetate
COC(=O)C(=O)Cl
This item is not returnable. View return policy
5781-53-3
C3H3ClO3
122.51
MFCD00000705
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
79846
methyl 2-chloro-2-oxoacetate
COC(=O)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Density 1.3300g/mL
Boiling Point 118°C to 120°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCO2CH3
Refractive Index 1.4179 to 1.4199
Quantity 25 mL
Beilstein 02, 541
Specific Gravity 1.33
Solubility Information Solubility in water: decomposes
Formula Weight 122.51
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl oxalyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl oxalyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'