missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methylenediphosphonic acid, 97%

Brand:  Alfa Aesar™ 030279.03

Code : NEW

Additional Details : CAS Number : 1984-15-2 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11337248

  • £20.20 / 1g


Estimated Shipment date
from Supplier 19-06-2018
Add to basket

Specifications

Chemical Identifiers

Methylenediphosphonic acid
176
UN3261
14,5792
1984-15-2
MFCD00002139
1708494
Bis(dihydroxyphosphoryl)methane; Medronic acid

Specifications

Methylenediphosphonic acid
Crystalline
176
UN3261
14,5792
202° to 204°C
Odorless
1984-15-2
97%
MFCD00002139
1708494
Bis(dihydroxyphosphoryl)methane; Medronic acid
1g
Videos
SDS