missing translation for 'onlineSavingsMsg'
Learn More

Methylenecyclohexane, 98%, Thermo Scientific Chemicals

Catalog Number p-3758396
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
1192-37-6
C7H12
96.17
MFCD00001661
YULMNMJFAZWLLN-UHFFFAOYSA-N
methylenecyclohexane, cyclohexane, methylene, 1-methylenecyclohexane, methylene-cyclohexane, cyclohexylmethyl, methylcyclohexyl, acmc-209a1w, methylene cyclohexane
14502
methylidenecyclohexane
C=C1CCCCC1
1192-37-6
C7H12
96.17
MFCD00001661
YULMNMJFAZWLLN-UHFFFAOYSA-N
methylenecyclohexane, cyclohexane, methylene, 1-methylenecyclohexane, methylene-cyclohexane, cyclohexylmethyl, methylcyclohexyl, acmc-209a1w, methylene cyclohexane
14502
methylidenecyclohexane
C=C1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8000g/mL
Boiling Point 102.0°C to 103.0°C
Flash Point −006°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(=CH2)
Refractive Index 1.4465 to 1.4505
Quantity 1 g
Beilstein 05, 69
Specific Gravity 0.8
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methylenecyclohexane
Show More Show Less
compliance-icons
Product Identifier
  • Methylenecyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'