missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methylenebis(3-thiopropionic acid), 97%

Brand:  Alfa Aesar™ A10278.14

Code : NEW

Additional Details : CAS Number : 4265-57-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11414013

  • £55.01 / 25g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Methylenebis (3-thiopropionic acid)
CH2(SCH2CH2CO2H)2
MFCD00020522
4265-57-0
224.29
Methylenebis(3-mercaptopropionic acid)

Specifications

Methylenebis (3-thiopropionic acid)
97%
224.29
Methylenebis(3-mercaptopropionic acid)
25g
4265-57-0
CH2(SCH2CH2CO2H)2
MFCD00020522
141° to 143°C
Stench
Videos
SDS