missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, Low water, BakerDRY™, J.T. Baker™

Catalog Number p-5391002
212.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Formula Weight 84.93
Grade BakerDRY™
Quantity 1 L
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Specific Gravity 1L = 1.32kg
Chemical Name or Material Methylene Chloride, Low water
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'