missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-butyrolactone, 95%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3758400
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
547-65-9
C5H6O2
98.1
MFCD00005407
GSLDEZOOOSBFGP-UHFFFAOYSA-N
2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
68352
CHEBI:104120
3-methylideneoxolan-2-one
C=C1CCOC1=O
This item is not returnable. View return policy
547-65-9
C5H6O2
98.1
MFCD00005407
GSLDEZOOOSBFGP-UHFFFAOYSA-N
2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
68352
CHEBI:104120
3-methylideneoxolan-2-one
C=C1CCOC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow or Pink
Density 1.1200g/mL
Boiling Point 88°C to 92°C (12.0 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.475
Quantity 1 g
Fieser 11,80
Merck Index 15, 6134
Specific Gravity 1.12
Stabilizer 2, 6-di-tert.-butyl-p-cresol
Formula Weight 98.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material α-Methylene-γ-butyrolactone, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • alpha-Methylene-gamma-butyrolactone, stabilized
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'