missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue trihydrate, Thermo Scientific Chemicals

Catalog Number 15433529
96.90 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
7220-79-3
C16H24ClN3O3S
373.90
MFCD00150008
XQAXGZLFSSPBMK-UHFFFAOYSA-M
C.I. 52015; Basic Blue 9
104827
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
7220-79-3
C16H24ClN3O3S
373.90
MFCD00150008
XQAXGZLFSSPBMK-UHFFFAOYSA-M
C.I. 52015; Basic Blue 9
104827
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Sensitive stain for RNA and ribonuclease

Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

Solubility
Soluble in water, alcohol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Beilstein 3599847
Merck Index 14,6060
Solubility Information Soluble in water,alcohol and chloroform; Insoluble in ether.
Formula Weight 373.90 (319.86 Anhydrous)
Odor Odorless
Color Green
Quantity 250 g
Physical Form Crystalline Powder
Chemical Name or Material Methylene Blue trihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'