missing translation for 'onlineSavingsMsg'
Learn More

Methyl isothiocyanate, 97%, Acros Organics™

Brand:  Acros Organics 414470250

Code : 29

Additional Details : CAS Number : 556-61-6 Weight : 0.07500kg Transport : UN number : 2477 Chem class : 6.1 Pack group : I


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10249360

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Methyl isothiocyanate
97%
CH3NCS
MFCD00004818
97%
C2H3NS
73.12
MTC, Methyl mustard-oil

Specifications

Methyl isothiocyanate
1.0690g/mL
Glass bottle
Low Melting Solid
97%
CH3NCS
MFCD00004818
Pale yellow to yellow-orange
MTC, Methyl mustard-oil
25g
556-61-6
Authentic
1.069
97%
C2H3NS
73.12
15, 6164
117.0° to 120.0°C
30.0° to 36.0°C
32°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl isothiocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS