missing translation for 'onlineSavingsMsg'
Learn More

Methylecyclobutanecarboxylate, 98%, ACROS Organics™

Brand:  Acros Organics 365540250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 765-85-5 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10560944

  • £90.20 / 25g


Estimated Shipment
18-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Methylecyclobutanecarboxylate
765-85-5
98%
114.14
98%
≥97.5% (GC)
C6H10O2
MFCD00095142

Specifications

Methylecyclobutanecarboxylate
25g
765-85-5
Glass bottle
Crystalline Powder and Lumps
98%
114.14
White to beige
98%
Authentic
1.4260 to 1.4300 (20°C, 589nm)
≥97.5% (GC)
C6H10O2
MFCD00095142

Safety and Handling

Product Identifier
  • Methyl cyclobutanecarboxylate

Documentation

Technical Documents