missing translation for 'onlineSavingsMsg'
Learn More

Methylecyclobutanecarboxylate, 98%, ACROS Organics™

Brand:  Acros Organics 365540050

Code : 29

Additional Details : CAS Number : 765-85-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10662993

  • £28.60 / 5g


Estimated Shipment
14-06-2018
Add to basket

Specifications

Chemical Identifiers

Methylecyclobutanecarboxylate
765-85-5
98%
114.14
98%
≥97.5% (GC)
C6H10O2
MFCD00095142

Specifications

Methylecyclobutanecarboxylate
5g
765-85-5
Glass bottle
Crystalline Powder and Lumps
98%
114.14
White to beige
98%
Authentic
1.4260 to 1.4300 (20°C, 589nm)
≥97.5% (GC)
C6H10O2
MFCD00095142
Videos
SDS